Learn More
Promega pBR322 Vector
Carries the genes for tetracycline and ampicillin resistance
Supplier: Promega D1511
Description
- Plasmid Subcloning Vector that Carries Genes for Tetracycline and Ampicillin Resistance
- Supplied at a concentration of 1mg/ml
Spécifications
| 10mM Tris HCl (pH 7.5), 1mM EDTA | |
| 1 μg/μL | |
| pBR322 DNA vector |
| -30°C to -10°C | |
| 10 μg | |
| Carries the genes for Tetracycline and Ampicillin resistance and DNA digests are used as molecular weight size markers in Gel analysis of Nucleic Acids |