Learn More
Promega NAD/NADH-Glo™ Assay
The NAD/NADH-Glo™ Assay is a bioluminescent, homogeneous single-reagent-addition assay for detecting total oxidized and reduced nicotinamide adenine dinucleotides and determining their ratio in biological samples or in defined enzyme reactions.
Supplier: Promega G9071
Description
- Bioluminescent Assays to Detect NAD(P)+ and NAD(P)H Levels in Cells
- Homogeneous, single-reagent-addition assays
- Easily adapted for high throughput screening formats
Specifications
| 10 mL | |
| -20°C |