Learn More
Promega NAD/NADH-Glo™ Assay DFS Item
The NAD/NADH-Glo™ Assay is a bioluminescent, homogeneous single-reagent-addition assay for detecting total oxidized and reduced nicotinamide adenine dinucleotides and determining their ratio in biological samples or in defined enzyme reactions.
$1146.31 - $4751.27
Specifications
Content And Storage -20°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRG9071 DFS Item
View Documents
Promega
G9071
10 mL
Each for $1,146.31
Only null left
PRG9072 DFS Item
View Documents
Promega
G9072
50 mL
Each for $4,751.27
Only null left
Description
Description
  • Bioluminescent Assays to Detect NAD(P)+ and NAD(P)H Levels in Cells
  • Homogeneous, single-reagent-addition assays
  • Easily adapted for high throughput screening formats
Specifications
Specifications
-20°C
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'