Learn More
Promega MultiTox-Glo™ Multiplex Cytotoxicity Assay
The MultiTox-Glo™ Multiplex Cytotoxicity Assay is a fluorescent and luminescent assay that measures the relative number of live and dead cells in a population.
Supplier:  Promega G9270
Encompass
 View more versions of this product
Catalog No. PRG9270
Only null left
Includes: Assay buffer, AAF-Glo™ substrate, GF-AFC substrate
Description
Description
  • Measures Relative Number of Live and Dead Cells
  • Fluorescent and luminescent assay
  • Normalize data with a built-in internal control
  • Immediately identify more false-positives and false-negatives
Specifications
Chemical Identifiers
Fluorescence, Luminescence
-30°C to -10°C
Specifications
10 mL
Fluorescence, Luminescence
-30°C to -10°C
100 x 96-well, 400 x 384-well
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
  • Patent Pending.
  • U.S. Patents 6,602,677 and 7,241,584, Australian Patents 754312 and 785294 and other patents and patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.