missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega MultiTox-Glo™ Multiplex Cytotoxicity Assay
The MultiTox-Glo™ Multiplex Cytotoxicity Assay is a fluorescent and luminescent assay that measures the relative number of live and dead cells in a population.
$517.81 - $3,070.01
Specifications
| Detection Method | Fluorescence, Luminescence |
|---|---|
| Content And Storage | -30°C to -10°C |
Includes: Assay buffer, AAF-Glo™ substrate, GF-AFC substrate
| Catalog Number | Mfr. No. | Quantity | No. of Reactions | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | No. of Reactions | Price | Quantity & Availability | ||||
PRG9271
|
Promega
G9271 |
5 x 10 mL | 500 x 96-well and 2000 x 384-well |
Each for $1,727.37
|
|
||||
PRG9272
|
Promega
G9272 |
2 x 50 mL | 1000 x 96-well and 4000 x 384-well |
Each for $3,070.01
|
|
||||
PRG9270
|
Promega
G9270 |
10 mL | 100 x 96-well, 400 x 384-well |
Each for $517.81
|
|
||||
Description
- Measures Relative Number of Live and Dead Cells
- Fluorescent and luminescent assay
- Normalize data with a built-in internal control
- Immediately identify more false-positives and false-negatives
Specifications
| Fluorescence, Luminescence | |
| -30°C to -10°C |
- Patent Pending.
- U.S. Patents 6,602,677 and 7,241,584, Australian Patents 754312 and 785294 and other patents and patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.