missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega MagneSil™ Total RNA Mini-Isolation System
Description
Includes
MagneSil RNA Paramagnetic Particles, 20mL; DNase I, lyophilized, 4 x 1 vial; DNase Stop Solution, 26.5mL; MnCl2, 0.09M, 5mL; Nuclease-free water, 150mL; RNA Lysis Buffer, 100mL; Yellow Core Buffer, 25mL
Requires
Requires MagnaBot™ 96 Magnetic Separation Device (Mfr. No. PR-V8151) and 0.25 in. Foam Spacer (Mfr. No. PR-Z3301) for 96-well purification or MagnaBot 384 Magnetic Separation Device (Mfr. No. V8241) for 384-well purification.
- Fast, Automated RNA Isolation from Small Amounts of Cells, Tissue or Blood
- Liquid handler-compatible
- Concentrated RNA eluted in as little as 15μL
U.S. Patents 6,027,945 and 6,638,800; Australian Patent 732756 and other patents and patents pending.
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Format | 96-well Plate |
| For Use With (Application) | RT-PCR amplification and real time RT-PCR |
| Includes | RNA Lysis Buffer (RLA), 100mL, Magnesil™ RNA paramagnetic particles (PMPs) 20mL, DNase I (lyophilized) 4 vials, yellow core Buffer 25mL, MnCl2, 0.09M 5mL, DNase stop solution (concentrated) 26.5mL, Nuclease-free Water 150mL |
| Quantity | 4 x 384 Preps |