missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega M-MLV Reverse Transcriptase
M-MLV RT is an RNA-dependent DNA polymerase that can be used in cDNA synthesis with long messenger RNA templates (>5kb).
Supplier: Promega M1705
Description
- RNA-Dependent DNA Polymerase for cDNA Synthesis
- Enzyme supplied at a concentration of 200u/μL
Specifications
| 200 U/μL | |
| M-MLV Reverse Transcriptase | |
| 7.5 | |
| cDNA Synthesis | |
| Recombinant E. coli Strain |
| −30 to −10°C | |
| Liquid | |
| 50000U | |
| 5X reaction buffer (250mM Tris-HCl [pH 8.3 at 25°C], 375mM KCl, 15mM MgCl2, 50mM DTT), buffer also available separately | |
| Tris-HCl (pH 7.5 at 25°C) 20mM, NaCl 0.2M, EDTA 0.1mM, DTT 1mM, 0.01% Nonidet™ P40™ and Glycerol 50% |