missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega M-MLV Reverse Transcriptase
M-MLV RT is an RNA-dependent DNA polymerase that can be used in cDNA synthesis with long messenger RNA templates (>5kb).
$101.83 - $388.25
Specifications
| Content And Storage | −30 to −10°C |
|---|---|
| Form | Liquid |
| For Use With (Application) | cDNA Synthesis |
| Source | Recombinant E. coli Strain |
| Storage Buffer | Tris-HCl (pH 7.5 at 25°C) 20mM, NaCl 0.2M, EDTA 0.1mM, DTT 1mM, 0.01% Nonidet™ P40™ and Glycerol 50% |
Description
- RNA-Dependent DNA Polymerase for cDNA Synthesis
- Enzyme supplied at a concentration of 200u/μL
Specifications
| −30 to −10°C | |
| cDNA Synthesis | |
| Tris-HCl (pH 7.5 at 25°C) 20mM, NaCl 0.2M, EDTA 0.1mM, DTT 1mM, 0.01% Nonidet™ P40™ and Glycerol 50% |
| Liquid | |
| Recombinant E. coli Strain |