missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Luciferase SP6 Control DNA
Description
- Functional Controls for the TnT(R) Quick Coupled and TnT(R) Coupled Transcription/Translation Systems
- Contains luciferase gene under transcriptional control of an SP6 or T7 phage RNA polymerase promoter
- Includes a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene
- Easy to monitor control reactions with a luciferase assay
Specifications
Specifications
| Product Type | Luciferase SP6 Control DNA |
| Storage Buffer | TE Buffer |
| Content And Storage | -30°C to -10°C |
| Concentration | 0.5 mg/mL |
| Quantity | 20 μg |
| For Use With (Application) | TnT quick coupled and TnT toupled transcription/translation systems |