missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Kinase-Glo™ Max Luminescent Kinase Assay
The Kinase-Glo™ Luminescent Kinase Assays are homogeneous non-radioactive systems for determining the activity of purified kinases by quantifying the amount of ATP remaining in solution following a kinase reaction.
Supplier: Promega V6073
Description
- Monitors Activity of Purified Kinases by Quantifying ATP
- Luminescent signal is inversely proportional to the amount of kinase activity
- Three kit formats: Linear to 10microM (Kinase-Glo(R)); 100microM (Kinase-Glo(R) Plus) or 500microM (Kinase-Glo(R) Max) ATP
Chemical Identifiers
| -30°C to -10°C | |
| For determining the activity of purified Kinases by quantifying the amount of ATP | |
| For multiwell plates |
| Homogeneous, Non-radioactive Method | |
| Kinase-Glo Max Luminescent Kinase Assay |
Specifications
| 100 mL | |
| Homogeneous, Non-radioactive Method | |
| 1 x 100mL Kinase-Glo™ max Buffer, 1 x resuspend to 100mL Kinase-Glo™ max substrate | |
| For multiwell plates |
| -30°C to -10°C | |
| For determining the activity of purified Kinases by quantifying the amount of ATP | |
| Kinase-Glo Max Luminescent Kinase Assay |