missing translation for 'onlineSavingsMsg'
Learn More

Promega Kinase-Glo™ Max Luminescent Kinase Assay

Catalog No. p-2799790
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
10 x 10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV6072 10 x 10 mL
PRV6071 10 mL
PRV6073 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRV6072 Supplier Promega Supplier No. V6072
Only null left

The Kinase-Glo™ Luminescent Kinase Assays are homogeneous non-radioactive systems for determining the activity of purified kinases by quantifying the amount of ATP remaining in solution following a kinase reaction.

  • Monitors Activity of Purified Kinases by Quantifying ATP
  • Luminescent signal is inversely proportional to the amount of kinase activity
  • Three kit formats: Linear to 10microM (Kinase-Glo(R)); 100microM (Kinase-Glo(R) Plus) or 500microM (Kinase-Glo(R) Max) ATP

  1. U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
  2. The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024, 5,674,713 and 5,700,673.

Specifications

Quantity 10 x 10 mL
Content And Storage -30°C to -10°C
Detection Method Homogeneous, Non-radioactive Method
For Use With (Application) For determining the activity of purified Kinases by quantifying the amount of ATP
Product Type Kinase-Glo Max Luminescent Kinase Assay
For Use With (Equipment) For multiwell plates