missing translation for 'onlineSavingsMsg'
Learn More

FuGENE™ HD Transfection Reagent

Catalog No. p-3853213
Encompass
Change view
Click to view available options
Quantity:
1 mL
5 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE2311 1 mL
PRE2312 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRE2311 Supplier FuGENE™ Supplier No. E2311
Only null left

FuGENE HD is a novel, nonliposomal reagent for transfecting DNA into a variety of cell lines with high efficiency and low toxicity.

  • Better Transfection for Challenging Cell Lines
  • Difficult-to-transfect cell lines
  • Transfection of primary cells
  • Protein and virus production

Specifications

Content And Storage 2°C to 10°C
For Use With (Application) Transfect DNA into a wide variety of cell lines with high efficiency and low toxicity, for use with luciferase assays, providing more expression and sensitive results
Quantity 1 mL

FuGENE™ HD is sold with a Limited Use Label License (LULL). You shall use this Product in accordance with all applicable laws, rules and regulations. You may not attempt to reverse engineer this Product. If you are not willing to accept these conditions, the distributor from whom the Product was purchased will accept the return of the unopened Product and provide you with a full refund. However, in the event this Product is opened, you agree to be bound by the conditions of this limited use statement.