missing translation for 'onlineSavingsMsg'
Learn More

Promega ECL Western Blotting Substrate

Catalog No. PRW1001
Encompass
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRW1001 250 mL
PRW1015 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRW1001 Supplier Promega Supplier No. W1001

The ECL Western Blotting Substrate is a highly sensitive non-radioactive, enhanced luminol-based chemiluminescent substrate for the detection of horseradish peroxidase (HRP) conjugates on immunoblots.

  • Chemiluminescent Substrate to Detect Horseradish Peroxidase (HRP) Conjugates on Immunoblots
  • Detect picogram levels of protein with minimal background
  • Non-radioactive
  • No optimization required; easily switch from other entry-level ECL substrates

Specifications

Product Type ECL Western Blotting Substrate
Quantity 250 mL
For Use With (Application) Detects picogram amounts of Antigen and presence of HRP, blots can be repeteadly exposed to X-ray
Storage Requirements 2°C to 10°C