missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat Anti-Mouse IgG (H+L), AP, Secondary Antibody, Promega
Ideal for use in protein detection in Western Blots and ELISAs.
Supplier: Promega Corporation S3721
Description
- Ready-to-Use Formulation: No need to dissolve the antibody
- Applications:
- Western blotting
- Enzyme-linked immunosorbent assay (ELISA)
- Dot blotting
Specifications
| Dot Blot, Western Blot | |
| Alkaline Phosphatase (AP) | |
| Goat | |
| RUO | |
| Store Anti-Mouse, Anti-Human, Anti-Rabbit (H+L) (Fc), AP conjugate antibodies at 2° to 10°C. Store Donkey Anti-goat IgG, AP at -30° to -10°C. |
| Polyclonal | |
| Anti-Mouse IgG (H+L) | |
| 100 μL | |
| Secondary | |
| Liquid |