Learn More
Goat Anti-Mouse IgG (H+L), HRP, Secondary Antibody, Promega DFS Item
Ideal for use in protein detection in Western Blots and ELISAs.
Supplier:  Promega Corporation W4021
Catalog No. PR-W4021
Only null left
Explore available promotions
Description
Description
  • Ready-to-Use Formulation: No need to dissolve the antibody
  • Applications:
  • Western blotting
  • Enzyme-linked immunosorbent assay (ELISA)
  • Dot blotting
Specifications
Specifications
Dot Blot, Western Blot
HRP
Goat
RUO
Store the unopened product at -20°C. Store opened product at 2 to 10°C. (Should be good at this temp. until expiration date.)
Polyclonal
Anti-Mouse IgG (H+L)
300 μL
Secondary
Liquid
Product Suggestions
Product Suggestions