missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat Anti-Rabbit IgG (H+L), HRP, Secondary Antibody, Promega
Ideal for use in protein detection in Western Blots and ELISAs.
Supplier: Promega Corporation W4011
Description
- Ready-to-Use Formulation: No need to dissolve the antibody
- Applications:
- Western blotting
- Enzyme-linked immunosorbent assay (ELISA)
- Dot blotting
Specifications
| Dot Blot, Western Blot | |
| HRP | |
| Goat | |
| RUO | |
| Store the unopened product at â20°C. Store opened Anti-Human IgG (H+L), HRP Conjugate, Anti-Mouse IgG (H+L), HRP Conjugate and Anti-Rabbit IgG (H+L), HRP Conjugate at 4°C. |
| Polyclonal | |
| Anti-Rabbit IgG (H+L) | |
| 300 μL | |
| Secondary | |
| Liquid |