missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega CellTiter 96™ AQueous One Solution Cell Proliferation Assay (MTS)
Homogeneous, colorimetric cell viability assay. Requires a 1- to 4-hour incubation.
$204.23 - $2,304.48
Specifications
| For Use With (Application) | Cytotoxicity |
|---|---|
| Format | Multiwell Plates |
| Content And Storage | -30°C to -10°C |
Description
- A Colorimetric Method for Determining the Number of Viable Cells
- Simple add-mix-measure format
- Non-radioactive assay
Specifications
| Cytotoxicity | |
| Multiwell Plates | |
| -30°C to -10°C |