missing translation for 'onlineSavingsMsg'
Learn More

Promega CellTiter 96™ AQueous One Solution Cell Proliferation Assay (MTS)

Catalog No. PRG3582
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
200 Assays
5000 Assays
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG3582 200 Assays
PRG3580 1000 Assays
PRG3581 5000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG3582 Supplier Promega Supplier No. G3582

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Homogeneous, colorimetric cell viability assay. Requires a 1- to 4-hour incubation.

  • A Colorimetric Method for Determining the Number of Viable Cells
  • Simple add-mix-measure format
  • Non-radioactive assay

Specifications

Quantity 200 Assays
Format Multiwell Plates
Content And Storage -30°C to -10°C
For Use With (Application) Cytotoxicity