missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega CaspACE™ FITC-VAD-FMK In Situ Marker
CaspACE FITC-VAD-FMK In Situ Marker is a fluorescent analog of the pan caspase inhibitor Z-VAD-FMK that is used to assay for caspase activity in situ.
$669.35 - $1,383.48
Specifications
| Content And Storage | -30°C to -10°C |
|---|---|
| For Use With (Application) | Fluorescence Detection |
Description
- A Fluorescent Pan-Caspase Inhibitor That Detects Caspase Activity
- Quick, single-reagent protocol
- No preparation of cell extracts needed
- Amenable to a variety of detection methods and high throughput applications
Specifications
| -30°C to -10°C | |
| Fluorescence Detection |