missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega CaspACE™ FITC-VAD-FMK In Situ Marker
Description
- A Fluorescent Pan-Caspase Inhibitor That Detects Caspase Activity
- Quick, single-reagent protocol
- No preparation of cell extracts needed
- Amenable to a variety of detection methods and high throughput applications
Specifications
Specifications
| Quantity | 50 μL |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | Fluorescence Detection |