missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Bovine Serum Albumin, Acetylated
Used as an enzyme stabilizer or as a carrier protein. Acetylated to be nuclease-free.
$46.63 - $46.63
Specifications
| Name | BSA Acetylated |
|---|---|
| Source | Bovine Serum |
| Storage Requirements | −30 to −10°C |
| Form | Liquid |
Description
- Quality tested and certified free of DNase and RNase activity
Specifications
| BSA Acetylated | |
| Bovine Serum | |
| −30 to −10°C | |
| Liquid |