missing translation for 'onlineSavingsMsg'
Learn More

Promega Bovine Serum Albumin, Acetylated

Catalog No. PRR3961
Encompass
Change view
Click to view available options
Quantity:
1 mL
Concentration:
10mg/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
PRR3961 1 mL 10mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRR3961 Supplier Promega Supplier No. R3961

Used as an enzyme stabilizer or as a carrier protein. Acetylated to be nuclease-free.

  • Quality tested and certified free of DNase and RNase activity

Specifications

Concentration 10mg/mL
Name BSA Acetylated
Quantity 1 mL
Source Bovine Serum
Storage Requirements −30 to −10°C
Form Liquid