Learn More
Promega Blot Qualified BSA DFS Item
BSA (bovine serum albumin) has been tested and qualified for optimum performance in immunoblotting applications with alkaline phosphatase antibody conjugates.
Supplier:  Promega W3841
Catalog No. PR-W3841
Only null left
Explore available promotions
Description
Description
  • Blot-Qualified BSA and Tween(R) 20 for Use as Blocking Agents for Antibody Detection
  • Blot-Qualified BSA saturates excess sites of antibody binding on membranes for AP antibody conjugates
  • Tween(R) 20 (100%) is used in immunoscreening to dissociate nonspecific interactions with an antibody probe
Specifications
Specifications
Immunoblotting
10 g
Powder
Blot Qualified BSA
15 to 30°C
Product Suggestions
Product Suggestions