missing translation for 'onlineSavingsMsg'
Learn More

Promega BCIP/NBT Color Development Substrate

Catalog No. PRS3771
Encompass
Change view
Click to view available options
Quantity:
1.25 or 2.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRS3771 1.25 or 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRS3771 Supplier Promega Supplier No. S3771
Only null left

For colorimetric detection of alkaline phosphatase activity.

  • Quality tested and qualified for use in blotting

Specifications

Formulation BCIP 50mg/mL in 100% Dimethylformamide, NBT 50mg/mL in 70% Dimethylformamide
For Use With (Application) For colorimetric detection of Alkaline Phosphatase activity
Sample Volume 1.25 or 2.5mL
Content And Storage −30 to −10°C
Product Type BCIP/NBT color development Substrate
Quantity 1.25 or 2.5 mL