missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega BacTiter-Glo™ Microbial Cell Viability Assay Kit
Description
Includes
BacTiter-Glo Buffer; BacTiter-Glo Substrate (lyophilized)
- A Luminescent Assay to Measure the Number of Viable Microbial Cells in Culture
- A homogeneous, single reagent assay
- Offers increased sensitivity, fast results
- U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
Specifications
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | Antimicrobial Compound Activity, Antimicrobial Compounds Screening, Viability Assay, Rapid Bacterial Quantitation, Rapid ATP Detection |
| For Use With (Equipment) | Microplate Reader |
| Includes | 1 x 100mL BacTiter-Glo™ Buffer, 10 x 1000mL BacTiter-Glo™ substrate (lyophilized) |
| Quantity | 10 x 100mL |
| Sufficient For | 10,000 Assays at 100μL/assay in 96-well plates, 40,000 assays at 25μL/assay in 384-well plates |