Learn More
Promega Bacterial Strain JM109
Description
- Useful Host for Transforming Plasmids Capable of Blue/White Screening
- Reliable strain that grows well and is transformed efficiently
- Useful for cloning, single-stranded DNA production, and blue/white screening
- Yield and quality of plasmid DNA improved with the endonuclease A- mutation
Specifications
Specifications
| Content And Storage | <-65°C |
| Quantity | 500 μL |