missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proguanil hydrochloride, 97%
CAS: 637-32-1 | C11H16ClN5·HCl | 290.19 g/mol
$156.63 - $156.63
Chemical Identifiers
| CAS | 637-32-1 |
|---|---|
| Molecular Formula | C11H16ClN5·HCl |
| Molecular Weight (g/mol) | 290.19 |
| InChI Key | SARMGXPVOFNNNG-UHFFFAOYSA-N |
| Synonym | proguanil hydrochloride, diguanyl, paludrine, proguanil hcl, chlorguanide hydrochloride, chloroquanil, bigumalum, chloroguanide hydrochloride, tirian hydrochloride, palusil hydrochloride |
| PubChem CID | 9570076 |
| IUPAC Name | (1E)-1-[amino-(4-chloroanilino)methylidene]-2-propan-2-ylguanidine;hydrochloride |
| SMILES | CC(C)N=C(N)N=C(N)NC1=CC=C(C=C1)Cl.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC461430250
|
Thermo Scientific Chemicals
461430250 |
25 mg |
Each for $156.63
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 637-32-1 | |
| 290.19 | |
| proguanil hydrochloride, diguanyl, paludrine, proguanil hcl, chlorguanide hydrochloride, chloroquanil, bigumalum, chloroguanide hydrochloride, tirian hydrochloride, palusil hydrochloride | |
| (1E)-1-[amino-(4-chloroanilino)methylidene]-2-propan-2-ylguanidine;hydrochloride |
| C11H16ClN5·HCl | |
| SARMGXPVOFNNNG-UHFFFAOYSA-N | |
| 9570076 | |
| CC(C)N=C(N)N=C(N)NC1=CC=C(C=C1)Cl.Cl |
Specifications
| 245.0°C to 250.0°C | |
| 25 mg | |
| 96% min. (HPLC) | |
| proguanil hydrochloride, diguanyl, paludrine, proguanil hcl, chlorguanide hydrochloride, chloroquanil, bigumalum, chloroguanide hydrochloride, tirian hydrochloride, palusil hydrochloride | |
| CC(C)N=C(N)N=C(N)NC1=CC=C(C=C1)Cl.Cl | |
| 290.19 | |
| 290.19 | |
| Proguanil hydrochloride |
| 637-32-1 | |
| Authentic | |
| C11H16ClN5·HCl | |
| SARMGXPVOFNNNG-UHFFFAOYSA-N | |
| (1E)-1-[amino-(4-chloroanilino)methylidene]-2-propan-2-ylguanidine;hydrochloride | |
| 9570076 | |
| 97% |
Safety and Handling
Toxic if swallowed
Acute toxicity (category 3)
EINECSNumber : 211-283-7
RUO – Research Use Only