missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 57-83-0 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available. |
| Linear Formula | C21H30O2 |
| MDL Number | MFCD00003658 |
| Melting Point | 128°C to 132°C |
| Synonym | 4-Pregnene-3,20-dione |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 314.46 |
| Grade | Certified Reference Material |