Learn More
Applied Biosystems™ ProFlex™ PCR System + Plastics Packages, 1 x 96-well
The ProFlex PCR System combines the reliability and performance you’ve come to expect from Applied Biosystems instruments with the flexible configuration and control features that adapt to your research needs.
Supplier: Applied Biosystems™ A41184
Description
The ProFlex PCR System combines the reliability and performance you’ve come to expect from Applied Biosystems instruments with the flexible configuration and control features that adapt to your research needs. This package includes a system plus plastics (MicroAmp 8-Tube Strips with Attached Domed Caps or MicroAmp EnduraPlates and MicroAmp Clear Adhesive Films).
Proflex PCR System
• Interchange 96-well 0.2-mL block with another block type if throughput needs change
• Access the system remotely (and conveniently) through mobile app
• Program instrument in seconds with simple-to-use touchscreen interface
• Simulate old instrument with Thermal Simulation modes
• Optimize PCR quickly and efficiently with better-than-gradient VeriFlex blocks
MicroAmp 8-Tube Strips with Attached Domed Caps
MicroAmp 8-Tube Strips with Attached Domed Caps feature individually attached domed caps, dual large end-tabs, and graduated 20-μL markers on each tube to help prevent cross-contamination, pipetting errors, and sample identification errors.
• Strip of eight tubes with attached domed caps that open and close independently to prevent contamination and evaporation
• Angled caps that provide efficient tube strip positioning in racks and thermal cycler blocks
• Etched A-H letter labeling on individual tubes visible when caps are open or closed
• Dual end tabs for proper labeling and easy handling
• Graduated 20-μL measuring markers on each tube for visual volume verification
MicroAmp EnduraPlates
MicroAmp EnduraPlates offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. They are a solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments. They are validated for use with Applied Biosystems PCR thermal cyclers for optimal fit and performance.
• Optimized for performance with Applied Biosystems instruments
• Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
• Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling
• Available in multiple colors for easy organization of laboratory assays
• Excellent mechanical stability and flatness for your complete workflow needs
MicroAmp Clear Adhesive Films
MicroAmp Clear Adhesive Films create a secure seal across an entire microplate to prevent evaporation and well-to-well contamination.
• Adhesive provides tight seal over wells to prevent leaks and evaporation.
• Easy to apply and remove
• Available as Clear (economical and sticky) or Optical (ready for real-time PCR and non-tacky)
Chemical Identifiers
| ±0.25°C (35°C to 99.9°C) | |
| <0.5°C (20 sec. after reaching 95°C) | |
| 22 x 13 x 10.6 in. (56.5 x 33 x 27.2 cm) | |
| 100/240 V | |
| PCR System | |
| 0°C to 100°C | |
| 6°C/sec. (Block), 4.4°C/sec. (Sample) | |
| 41 lb. (18.8 kg) |
| Manual Protocols | |
| Fast, Standard | |
| 96-well, 0.2 mL | |
| 8.4 in. Color TFT LCD Touchscreen | |
| Limited Warranty | |
| USB, WiFi, Cloud, Ethernet | |
| 1 package | |
| 96-well, 6-Zone VeriFlex Block |
Specifications
| ±0.25°C (35°C to 99.9°C) | |
| Manual Protocols | |
| Fast, Standard | |
| 96-well, 0.2 mL | |
| 8.4 in. Color TFT LCD Touchscreen | |
| Limited Warranty | |
| USB, WiFi, Cloud, Ethernet | |
| Includes 5 x MicroAmp EnduraPlate Optical 96-well Clear Reaction Plates with Barcode (100 plates total) | |
| 41 lb. (18.8 kg) |
| • ProFlex 1 x 96-well PCR System • 5 x MicroAmp EnduraPlate Optical 96-well Clear Reaction Plates with Barcode (100 plates total) • 1 x MicroAmp Clear Adhesive Films (100 films) |
|
| <0.5°C (20 sec. after reaching 95°C) | |
| 22 x 13 x 10.6 in. (56.5 x 33 x 27.2 cm) | |
| 100/240 V | |
| PCR System | |
| 0°C to 100°C | |
| 6°C/sec. (Block), 4.4°C/sec. (Sample) | |
| 1 package | |
| 96-well, 6-Zone VeriFlex Block |