Learn More
Applied Biosystems™ ProFlex™ PCR System + Plastics Package, 2x96-well, EnduraPlates
Description
- Interchange the 96-well 0.2 mL block with another block type if your throughput needs change
- Access the system remotely (and conveniently) through a mobile app
- Program the instrument in seconds with a simple-to-use touchscreen interface
- Simulate your old instrument with Thermal Simulation modes
- Optimize your PCR quickly and efficiently with better-than-gradient VeriFlex blocks
- Optimized for performance with Applied Biosystems instruments
- Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
- Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling
- Excellent mechanical stability and flatness for your complete workflow needs
Specifications
Specifications
| Thermal Accuracy | ±0.25°C (35°C to 99.9°C) |
| Content And Storage | • ProFlex 2 x 96-well PCR System • 5 x MicroAmp EnduraPlate Optical 96-well Clear Reaction Plates with Barcode (100 plates total) • 1 x MicroAmp Clear Adhesive Films (100 films) |
| High-throughput Compatibility | Manual Protocols |
| Thermal Uniformity | <0.5°C (20 sec. after reaching 95°C) |
| Reaction Speed | Fast, Standard |
| Dimensions | 22 x 13 x 10.6 in. (56.5 x 33 x 27.2 cm) |
| Capacity | 192-well, 0.2 mL |
| Voltage | 100/240 V |
| Display Type | 8.4 in. Color TFT LCD Touchscreen |
| Product Type | PCR System |
| Show More |
Frequently Asked Questions (FAQs)
There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.
As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.
Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf
For Research Use Only. Not for use in diagnostic procedures.