Learn More
Applied Biosystems™ ProFlex™ PCR System, 2 x Flat
Description
The Veriti 96-well fast thermal cycler delivers the proven reliability of Applied Biosystems instruments. With the added control of VeriFlex blocks, you have six independent temperature blocks that provide precise control over your PCR optimization. Additionally, the color touchscreen simplifies the setup and use of the Veriti 96-well fast thermal cycler. Optional setups for fast or standard PCR methods provide flexibility to shorten your PCR cycling times. The Veriti 96-well fast thermal cycler comes with a 0.1-mL block configuration that saves time and reagent consumption.
Features of the Veriti 96-well fast thermal cycler include:
- Innovative VeriFlex blocks allow for precise PCR optimization or to run up to six independent assays
- Six temperature zones for convenient PCR optimization
- Standard- and fast-enabled cycling to address current and future PCR needs
- Easy-to-use 6.5-inch VGA touchscreen
- Convenient protocol transfer from one Veriti thermal cycler to another using USB
- Reduced PCR cycling time when using faster ramp rates
- Optional VeritiLink Remote Management Software allows remote management of more than 50 cyclers
VeriFlex blocks enhanced PCR functionality
Separate peltier blocks provide maximal versatility, flexibility, and offer two key benefits:
- Optimization—each block can be set with a specific temperature for precise control over PCR optimization
- More experiments—six peltier blocks can be used to run different annealing temperatures in the same run
Control at your fingertips
The powerful, yet simple to operate user interface on the Veriti system is driven by the 6.5-inch (16.51-cm) VGA color touchscreen. The large screen allows for easy viewing of temperature profiles, and large navigation buttons put programming at your fingertips. Setup and navigation of the Veriti Thermal Cycler does not require the use of a stylus or mouse.
Fast setup, fast results
The Veriti system has two options for method navigation. For quick setup, you may select one of the pre-programmed methods to run. If you prefer to program your own methods, simply touch the step you would like to setup and then use the keypad to enter thermal cycling steps. After completing a setup, you have the choice of saving the method to the instrument or to a USB memory stick.
Control your time, go fast when you want
With the Veriti Thermal Cycler you get two thermal cyclers in one: a standard thermal cycler with its better than gradient feature and a fast-enabled thermal cycler. So whether you have a current need to run fast PCR and shorten your PCR cycling times, or a possible future application need for fast PCR, the Veriti Thermal Cycler provides you with the flexibility to go fast when you want.
Applied Biosystems PCR plastics
Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for any experiment. To find the best match please visit our Plastics Selection Guide.
Specifications
Specifications
| Block Format | Interchangeable |
| Thermal Accuracy | ±0.25°C (35°C to 99.9°C) |
| High-throughput Compatibility | High-throughput Compatible |
| Thermal Uniformity | <0.5°C (20 sec. after reaching 95°C) |
| Reaction Speed | Fast, Standard |
| Dimensions | 22 x 13 x 10.6 in. (56.5 x 33 x 27 cm) |
| Capacity | 8 x 3072 OpenArray Plate, 24 x 20K Chip |
| Voltage | 100/240 V |
| Product Line | ProFlex |
| Display Type | 8.4 in. Color TFT LCD Touchscreen |
| Show More |
Frequently Asked Questions (FAQs)
There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.
As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.
Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf
For Research Use Only. Not for use in diagnostic procedures.