Learn More
Applied Biosystems™ ProFlex™ PCR System Extended Warranty Package, 2 x 96-well
Catalog No. A27938
Encompass
Change view
Click to view available options
Warranty:
3 years of total service coverage with instrument rapid exchange
5 years of total service coverage with 3 PMs
5 years of total service coverage with instrument rapid exchange
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Warranty
A27938 3 years of total service coverage with instrument rapid exchange
A27939 5 years of total service coverage with 3 PMs
A27937 5 years of total service coverage with instrument rapid exchange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A27938 Supplier Applied Biosystems™ Supplier No. A27938
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ProFlex™ PCR Systems are the latest in our line of Applied Biosystems™ thermal cyclers, providing the reliability and performance you’ve come to expect from Applied Biosystems instruments. This package includes the ProFlex PCR System and an extended warranty.
ProFlex™ PCR Systems are the latest in our line of Applied Biosystems™ thermal cyclers, providing the reliability and performance you've come to expect from Applied Biosystems instruments. By combining an extended warranty with your instrument purchase, you'll extend the life of your investment and receive the support you need to keep your instrument running at peak performance.
Each ProFlex PCR System Extended Warranty Package includes instrument configuration and control features, as well as extended warranty benefits, that adapt to your research needs:
  • Interchange block types if your throughput needs change
  • Access the system remotely (and conveniently) through a mobile app
  • Program the instrument in seconds with a simple-to-use touchscreen interface
  • Simulate your old instrument with Thermal Simulation modes
  • Receive fast repair from factory-trained and certified service engineers
  • Minimize instrument downtime by troubleshooting with experienced Technical Support teams
For detailed instrument information and specifications, see the ProFlex PCR System (Cat. No. 4484075).
Rapid Exchange (REX) Extended Warranty
Add additional warranty coverage to your existing manufacturer's limited warranty with the Rapid Exchange (REX) extended warranty. If your instrument needs repair, just call our Support Center and we'll send you a factory-certified refurbished replacement instrument within one business day from the time we determine a replacement is required. Instruments are typically received within 3 days of shipment and are yours to keep. All parts, labor, and shipping charges are included, and existing warranty contracts will be transferred to replacement units.
The Rapid Exchange extended warranty is ideal for customers with user-installable instruments that do not require asset tag consistency, and for those operating under critical timelines.
Certain exclusions apply. Contact your Service and Support Representative for details at service.sales@thermofisher.com.
Specifications
For Use With (Equipment) ProFlex™ PCR System
Product Line ProFlex
Warranty 3 years of total service coverage with instrument rapid exchange
Quantity System + 1 yr REX ext warranty
Warranty Services Not Included User-replaceable Parts, Planned Maintenance
Warranty Response Time Replacement instrument shipped in 1 business day and received within 3 days