missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ProFlex™ PCR System Extended Warranty Package, 2 x 384-well

Catalog No. A30757
Encompass
Change view
Click to view available options
Quantity:
System + 1 year ABRC Extended Warranty
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30757 System + 1 year ABRC Extended Warranty
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A30757 Supplier Applied Biosystems™ Supplier No. A30757

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ProFlex™ PCR Systems are the latest of Applied Biosystems thermal cyclers, providing the reliability and performance you've come to expect from Applied Biosystems instruments. By combining an extended warranty with your instrument purchase, you'll extend the life of your investment and receive the support you need to keep your instrument running at peak performance.

ProFlex™ PCR Systems are the latest in our line of Applied Biosystems™ thermal cyclers, providing the reliability and performance you've come to expect from Applied Biosystems instruments. By combining an extended warranty with your instrument purchase, you'll extend the life of your investment and receive the support you need to keep your instrument running at peak performance.

Each ProFlex PCR System Extended Warranty Package includes instrument configuration and control features, as well as extended warranty benefits, that adapt to your research needs:

  • Interchange block types if your throughput needs change
  • Access the system remotely (and conveniently) through a mobile app
  • Program the instrument in seconds with a simple-to-use touchscreen interface
  • Simulate your old instrument with Thermal Simulation modes
  • Receive fast repair from factory-trained and certified service engineers
  • Minimize instrument downtime by troubleshooting with experienced Technical Support teams

For detailed instrument information and specifications, see the ProFlex PCR System (Cat. No. 4484075).

AB Repair Center (ABRC) Extended Warranty

Add additional years of warranty coverage to your existing manufacturer's limited warranty with the AB Repair Center (ABRC) extended warranty. With ABRC support, factory-trained engineers will maintain and repair your instrument at our remote service center. Just mail us your instrument and we'll maintain or repair it and ship it back to you within 3 weeks. All parts, labor, and shipping charges are included.

The ABRC extended warranty is ideal for customers in regulated environments that must maintain asset tag consistency to comply with regulated protocols.

Certain exclusions apply. Contact your Service and Support Representative for details at service.sales@thermofisher.com.

Order Info

The Rapid Exchange extended warranty is ideal for customers with user-installable instruments that do not require asset tag consistency, and for those operating under critical timelines.

Certain exclusions apply. Contact your Service and Sup

Warranty

System and 1 year ABRC extended warranty

Specifications

Thermal Accuracy ±0.25°C (35°C to 99.9°C)
High-throughput Compatibility High-throughput Compatible
Thermal Uniformity <0.5°C (20 sec. after reaching 95°C)
Reaction Speed Fast, Standard
Dimensions 22 x 13 x 10.6 in. (56.5 x 33 x 27 cm)
Capacity 768 x 20 μL Tubes, 2 x 384-well Plate
Voltage 100/240 V
Product Line ProFlex
Display Type 8.4 in. Color TFT LCD Touchscreen
Product Type PCR System
Warranty 3 years of total service coverage
Thermal Range 0°C to 100°C
For Use With (Application) PCR
Programs Auto re-start (after Power Outages), Program Overwrite Protection
User Interface USB, WiFi, Cloud, Ethernet
Max. Ramp Rate 3°C/sec. (Block), 1.6°C/sec. (Sample)
Quantity System + 1 year ABRC Extended Warranty
Memory USB and On-board
Weight 45 lb. (20.4 kg)
Block Configurations 2 x 384-well Isothermal Blocks
Show More Show Less
What is the optimal ramp rate for my thermal cycler?

There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.

As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.

Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf

For Research Use Only. Not for use in diagnostic procedures.