missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ ProFlex™ 9015F(x) ANSI/ISO-Certified Anti-Vibration Gloves with DIR Protection
Full-length AVC™ palm padding reduces vibration and palm impact, while molded TPR armor on the knuckles protects the back of the hand.
Supplier: Ergodyne™ 17305
Description
- ANSI S2.73-2014/ISO 10819:2013-certified anti-vibration protection
- Full-length AVC™ palm padding reduces vibration and palm impact
- Molded TPR armor on the knuckles and fingers protects the back of the hand
- Palm coating enhances grip for a secure hold on tools and equipment
- Hex stretch knit, breathable spandex construction
- Low-profile molded hook & loop closure
- ID space for user personalization
- Reinforced thumb saddle
Specifications
| TPR Padding | |
| Black | |
| Spandex | |
| X-Large | |
| Assembly, Demolition, Heavy Construction, and Oil/Gas Refining | |
| AVC™ Padding |
| ANSI S2.73-2014/ISO 10819:2013 | |
| Specific | |
| Anti-vibration Gloves | |
| EN388:2121X | |
| Hook and Loop | |
| ProFlex™ 9015F(x) |