missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 9015F(x) ANSI/ISO-Certified Anti-Vibration Gloves with DIR Protection

Catalog No. p-7163138
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820148 Large
19820149 X-Large
19820150 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19820148 Supplier Ergodyne™ Supplier No. 17304
Only null left

Full-length AVC™ palm padding reduces vibration and palm impact, while molded TPR armor on the knuckles protects the back of the hand.

  • ANSI S2.73-2014/ISO 10819:2013-certified anti-vibration protection
  • Full-length AVC™ palm padding reduces vibration and palm impact
  • Molded TPR armor on the knuckles and fingers protects the back of the hand
  • Palm coating enhances grip for a secure hold on tools and equipment
  • Hex stretch knit, breathable spandex construction
  • Low-profile molded hook & loop closure
  • ID space for user personalization
  • Reinforced thumb saddle

Specifications

Back Material TPR Padding
Certifications/Compliance ANSI S2.73-2014/ISO 10819:2013
Color Black
Hand Specific
Material Spandex
Product Type Anti-vibration Gloves
Size Large
EN388 Standard EN388:2121X
For Use With (Application) Assembly, Demolition, Heavy Construction, and Oil/Gas Refining
Glove Cuff Style Hook and Loop
Palm Type AVC™ Padding
Series ProFlex™ 9015F(x)
Show More Show Less