Learn More
Ergodyne™ ProFlex™ 2000SF High-Performance Spandex Back Support Brace
ProFlex™ 2000SF High-Performance Spandex Back Support is designed to remind workers of proper lifting techniques while moving stress away from the spine for back pain relief.
Supplier: Ergodyne™ 11281
Description
- Non-conductive Sticky Fingers™ stays for long wear
- Innovative Power Panel Positioners™ ensure the tension bands stay close by
- Unique V-shaped design
- 840D spandex body material
- Adjustable, detachable suspenders keep back brace in place
- Waist size ranges from 25 to 58 in. (64 to 147 cm)
- Machine wash cold, air dry
Specifications
| Black | |
| X-Small | |
| Material Handling, Construction, Warehousing/Distribution, Maintenance | |
| 840D Spandex |