missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ ProFlex™ 2000SF High-Performance Spandex Back Support Brace
Description
- Non-conductive Sticky Fingers™ stays for long wear
- Innovative Power Panel Positioners™ ensure the tension bands stay close by
- Unique V-shaped design
- 840D spandex body material
- Adjustable, detachable suspenders keep back brace in place
- Waist size ranges from 25 to 58 in. (64 to 147 cm)
- Machine wash cold, air dry
Specifications
Specifications
| Color | Black |
| Size | X-Small |
| For Use With (Application) | Material Handling, Construction, Warehousing/Distribution, Maintenance |
| Material | 840D Spandex |