missing translation for 'onlineSavingsMsg'
Learn More

Proflavine Sulfate, Hydrate, Spectrum™ Chemical

Catalog No. 18612950
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612950 100 g Amber Glass Bottle
18612949 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18612950 Supplier Spectrum Chemical Mfg Cor Supplier No. P1429100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1811-28-5

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Molecular Weight (g/mol) 258.29
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Proflavine Sulfate, Hydrate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.