missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proflavine Sulfate, Hydrate, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 258.29 |
| Grade | Ungraded |
| Physical Form | Powder or Waxy Solid |
| Chemical Name or Material | Proflavine Sulfate, Hydrate |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.