missing translation for 'onlineSavingsMsg'
Learn More

Proflavine Hemisulfate Hydrate 97.0+%, TCI America™

Catalog No. p-7126011
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00735G 5 g
D007325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D00735G Supplier TCI America Supplier No. D00735G
Only null left

Chemical Identifiers

CAS 1811-28-5
Molecular Formula C13H15N3O5S
Molecular Weight (g/mol) 325.339
MDL Number MFCD00013150
InChI Key MSAVMVWXUVKHBI-UHFFFAOYSA-N
Synonym 3,6-Diaminoacridinium Monohydrogen Sulfate
PubChem CID 51062910
IUPAC Name acridine-3,6-diamine;sulfuric acid;hydrate
SMILES C1=CC(=CC2=NC3=C(C=CC(=C3)N)C=C21)N.O.OS(=O)(=O)O

Specifications

Quantity 5 g
Formula Weight 516.58
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Proflavine Hemisulfate Hydrate
RTECSNumber AR9065100
TSCA Yes