missing translation for 'onlineSavingsMsg'
Learn More

Gaylord Chemical Company, LLC PROCIPIENT™ (Dimethyl Sulfoxide USP, PhEur)
GREENER_CHOICE

Catalog No. p-200007203
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18614918 1 L
18614921 4 L
18614917 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18614918 Supplier Gaylord Chemical Company, LLC Supplier No. 5010430LT

Procipient™ DMSO is both an active pharmaceutical ingredient and a versatile excipient that is widely used in various pharmaceutical and biotechnology applications. Procipient is an extremely polar, water-soluble solvent with a minimum purity of 99.99%.

Procipient™ Characteristics:

  • Highly purified DMSO with a minimum purity of 99.99%
  • Water miscible/extremely polar solvent
  • Odorless, clear water-white liquid
  • Essentially non-toxic by all routes of administration
  • Chemically compatible with other excipients
  • Low viscosity (2.0 cP at 25°C)
  • Density similar to water
  • (1.0955 g / mL at 25°C)
  • Low volatility: 189°C boiling point

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.5°C (65.3°F)
Color Clear
Density 1.0955 g/mL (25°C)
Quantity 1 L
Flash Point 87°C (188.6°F) (Closed Cup)
Ignition Point 215°C (419°F)
Odor Odorless
Physical Form Liquid
Vapor Pressure 59.4 Pa at 20°C (68°F)
Viscosity 2.0 cP (25°C)
Boiling Point 189°C
Solubility Information Water Miscible, Extremely Polar Solvent
Percent Purity 99.99%
Grade USP
Chemical Name or Material Dimethyl Sulfoxide
Show More Show Less
Recommended Storage Store in a well-ventilated place. Keep cool.