missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Wash Buffer (10X)

Catalog No. p-200022107
Change view
Click to view available options
Quantity:
25 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125865 25 mL
EPX66666001 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501125865 Supplier Invitrogen™ Supplier No. EPX66666000
Only null left

ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates.

ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates. It is a simple mix of buffers, salts, and mild detergent.

ProcartaPlex Wash Buffer (10X) is included in our basic kits and off-the-shelf panels. Additionally, we offer it here as a stand-alone item.

Reported application

  • Multiplex immunoassay

Specifications

Product Type Buffer
Quantity 25 mL
For Use With (Equipment) Luminex™ Instruments

For Research Use Only.