Learn More
Invitrogen™ ProcartaPlex™ Wash Buffer (10X)
Description
ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates. It is a simple mix of buffers, salts, and mild detergent.
ProcartaPlex Wash Buffer (10X) is included in our basic kits and off-the-shelf panels. Additionally, we offer it here as a stand-alone item.
Reported application
- Multiplex immunoassay
Specifications
Specifications
| Product Type | Buffer |
| Quantity | 25 mL |
| For Use With (Equipment) | Luminex™ Instruments |
For Research Use Only.