missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex Rat Kidney Toxicity Panel 1 (5 plex)

Catalog No. 501125806
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125806 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501125806 Supplier Invitrogen™ Supplier No. EPX05030124901
Only null left

Includes

  • Rat Kidney Toxicity Panel 1 Standard Mix
  • Rat Kidney Toxicity Panel 1 Bead Mix
  • Rat Kidney Toxicity Panel 1 Detection Antibody Mix
  • Rat Kidney Toxicity Sample Diluent
  • Reading Buffer
  • Wash Buffer 10x
  • Streptavidin-PE
  • Universal Assay Buffer
  • Detection Antibody Diluent
  • PCR 8-Tube Strip
  • Adhesive Film
  • Greiner Microplate black 96 well

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • This kit cannot be combined with other ProcartaPlex Simplex or Multiplex Kits

Specifications

Quantity 96 Tests
Species Rat
For Use With (Application) Multiplex Immunoassay
Protein Subtype Calbindin, KIM-1, Osteopontin, Trefoil Factor 3, VEGF-A
Sample Type Urine
Sample Volume Urine: 25 μL
Synonym Convenience Panel