Learn More
Invitrogen™ eBioscience™ ProcartaPlex Rat IL-17A (CTLA-8) Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Supplier: Invitrogen™ EPX01A30635901
Includes:
- concentrated Magnetic Beads coated with monoclonal antibody to rat IL-17A
- rat IL-17A Standard (lyophilized)
- Biotin-Conjugate (50x) anti-rat IL-17A monoclonal antibody
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Specifications
| 96 Tests | |
| Multiplex Immunoassay | |
| Serum, Plasma: 25 μL; CCS: 50 μL |
| Rat | |
| Cell Culture Supernatant, Plasma, Serum | |
| Interleukin-17A, IL-17AA |