missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex Rat ICAM-1 Simplex
Description
Includes
- concentrated Magnetic Beads coated with monoclonal antibody to rat ICAM-1
- rat ICAM-1 Standard (lyophilized)
- Biotin-Conjugate (50x) anti-rat ICAM-1 monoclonal antibody
- Universal Assay Buffer 10x
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
- It is recommended to dilute sICAM-1 samples when measured in serum or plasma
- The recommended sample dilution factor is 1:200
Specifications
Specifications
| Quantity | 96 Tests |
| Species | Rat |
| For Use With (Application) | Multiplex Immunoassay |
| Protein Subtype | CD54 (ICAM-1) |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Sample Volume | Serum, Plasma: 25 μL; CCS: 50 μL |
| Synonym | Intercellular Adhesion Molecule 1, CD54 |