missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex Rat Antibody Isotyping Panel (6 plex)
Description
Includes
- Rat Isotyping Standard Mix
- Rat Isotyping Bead Mix
- Rat Isotyping Detection Antibody Mix
- Reading Buffer
- Wash Buffer 10x
- Universal Assay Buffer 10x
- Detection Antibody Diluent
- PCR 8-Tube Strip
- Adhesive Film
- Greiner Microplate black 96 well
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- Our assays are designed for use with the Luminex MAGPIX™, Luminex 100, Luminex 200, or Bio-Rad Bio-Plex analyzers
- This kit does not contain a Multiscreen Filter Plate
- It is recommended to dilute samples when measured in serum or plasma
- The recommended sample dilution factor is 1:40,000
Specifications
Specifications
| Quantity | 96 Tests |
| Species | Rat |
| For Use With (Application) | Multiplex Immunoassay |
| Protein Subtype | IgA, IgG1, IgG2a, IgG2b, IgG2c, IgM |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Sample Volume | Serum, Plasma: 25 μL; CCS: 50 μL |