missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex Porcine TNF alpha Simplex
Description
Includes
- concentrated Magnetic Beads coated with monoclonal antibody to porcine TNF alpha
- porcine TNF alpha Standard (lyophilized)
- Biotin-Conjugate (20x) anti-porcine TNF alpha monoclonal antibody
- ProcartaPlex™ assays are subject to the same stringent and robust development and manufacturing as are our extensively validated Platinum and Instant ELISA kits
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
Specifications
Specifications
| Quantity | 96 Tests |
| Species | Porcine |
| For Use With (Application) | Multiplex Immunoassay |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Sample Volume | Serum, Plasma: 25 μL; CCS: 50 μL |
| Synonym | Cachectin, Tumor necrosis factor alpha, DIF, TNFSF2 |