missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex Porcine IL-12/IL-23p40 Simplex

Catalog No. 501125786
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125786 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501125786 Supplier Invitrogen™ Supplier No. EPX01A62090901
Only null left

Includes

  • concentrated Magnetic Beads coated with monoclonal antibody to porcine IL-12 p40
  • porcine IL-12 p40 Standard (lyophilized)
  • Biotin-Conjugate (20x) anti-porcine IL-12p40 monoclonal antibody

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins

Specifications

Quantity 96 Tests
Species Porcine
For Use With (Application) Multiplex Immunoassay
Protein Subtype IL-12p40
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume Serum, Plasma: 25 μL; CCS: 50 μL
Synonym Interleukin-12 subunit beta, natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40