missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex NHP VCAM-1 Simplex

Catalog No. 501125734
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125734 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501125734 Supplier Invitrogen™ Supplier No. EPX01A40232901
Only null left

Includes

  • concentrated Magnetic Beads coated with monoclonal antibody to non-human primate sVCAM-1
  • non-human primate sVCAM-1 Standard (lyophilized)
  • Biotin-Conjugate (50x) anti-non-human primate sVCAM-1 monoclonal antibody
  • Universal Assay Buffer 10x

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • It is recommended to dilute sVCAM-1 samples when measured in serum or plasma
  • The recommended sample dilution factor is 1:200

Specifications

Quantity 96 Tests
Species Monkey
For Use With (Application) Multiplex Immunoassay
Protein Subtype sCD106 (sVCAM-1)
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume Serum, Plasma: 25 μL; CCS: 50 μL
Synonym Vascular cell adhesion protein 1, CD106 antigen